Learn More
Hidruro de aluminio y litio, sedimentos, 97 %, Thermo Scientific Chemicals
61.00€ - 999.00€
Identificadores de productos químicos
CAS | 16853-85-3 |
---|---|
Fórmula molecular | AlH4Li |
Peso molecular (g/mol) | 37.95 |
Número MDL | MFCD00011075 |
Clave InChI | OCZDCIYGECBNKL-UHFFFAOYSA-N |
Sinónimo | lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride |
PubChem CID | 21226445 |
SMILES | [Li+].[AlH4-] |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
11493800
|
Thermo Scientific Alfa Aesar
089643.14 |
25 g |
61.00€
25 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
11483800
|
Thermo Scientific Alfa Aesar
089643.22 |
100 g |
230.00€
100 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
11473800
|
Thermo Scientific Alfa Aesar
089643.36 |
500 g |
999.00€
500 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Lithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
16853-85-3 | |
37.95 | |
OCZDCIYGECBNKL-UHFFFAOYSA-N | |
21226445 |
AlH4Li | |
MFCD00011075 | |
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
[Li+].[AlH4-] |
Especificaciones
Lithium aluminum hydride | |
16853-85-3 | |
Sedimento | |
MFCD00011075 | |
14,349 | |
Soluble in water,ether and terahydrofuran. | |
[Li+].[AlH4-] | |
37.95 | |
37.95 | |
Odorless | |
0.917 g/mL |
>125°C (decomposition) | |
Blanco a gris | |
AlH4Li | |
UN1410 | |
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
OCZDCIYGECBNKL-UHFFFAOYSA-N | |
alumanuida de litio (1+) | |
21226445 | |
97% | |
Air and moisture sensitive |
Seguridad y manipulación
GHS H Statement
H260-H301-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314-H500
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM ALUMINUM HYDRIDE
missing translation for 'einecsNumber' : 240-877-9
missing translation for 'rtecsNumber' : BD0100000
missing translation for 'tsca' : Yes
Recommended Storage : Temperatura ambiente; almacenar bajo argón
RUO – Research Use Only