Learn More
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tube
Tienda Thermo Scientific Productos
Click to view available options
Volumen (métrico):
0,6 mL
1.5 mL
2 mL
Tamaño de la unidad:
paquete de 1000
paquete de 500
Descripción
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
Especificaciones
Especificaciones
| Esterilizables en autoclave | Autoclavable |
| Color | Natural |
| Material | Resina de polipropileno |
| Volumen (métrico) | 0,6 mL |
| Esterilidad | No estéril |
| Graduated | Sí |
| Tipo de tapón | Snap Cap |
| Fit for Purpose | PCR Ready |
| Forma del fondo | Conical |
| Forma | Cylindrical |
| Mostrar más |