Learn More
Loxiglumide, Tocris Bioscience™
CCK1 Receptor Antagonists
Marca: Tocris 3036/5
Detalles adicionales : Peso : 0.00970kg
Descripción
Loxiglumide is a CCK1 antagonist. Suppresses CCK-induced proliferation of acinar cells. Inhibits pancreatic secretion of digestive enzymes. Active in vivo.Especificaciones
107097-80-3 | |
4-[(3,4-Dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid | |
5 mg |