missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LRPAP1 (Human) Recombinant Protein
Human LRPAP1 full-length ORF (BAG35576.1, 1 a.a. - 357 a.a.) recombinant protein with GST tag at N-terminal.
Marca: Abnova™ H00004043-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Sequence: MAPRRVRSFLRGLPALLLLLLFLGPWPAASHGGKYSREKNQPKPSPKRESGEEFRMEKLNQLWEKAQRLHLPPVRLAELHADLKIQERDELAWKKLKLDGLDEDGEKEARLIRNLNVILAKYGLDGKKDARQVTSNSLSGTQEDGLDDPRLEKLWHKAKTSGKFSGEELDKLWREFLHHKEKVHEYNVLLETLSRTEEIHENVISPSDLSDIKGSVLHSRHTELKEKLRSINQGLDRLRRVSHQGYSTEAEFEEPRVIDLWDLAQSANLTDKELEAFREELKHFEAKIEKHKHYQKQLEIAHEKLRHAESVGDGERVSRSREKHALLEGRTKELGYTVKKHLQDLSGRISRARHNEL
Especificaciones
BAG35576.1 | |
low density lipoprotein receptor-related protein associated protein 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LRPAP1 | |
GST |
4043 | |
Wheat germ expression system | |
10 μg | |
A2MRAP, A2RAP, HBP44, MGC138272, MRAP, RAP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |