Learn More
Invitrogen™ LysoLight™ Antibody Labeling Kits
Description
LysoLight Antibody Labeling Kits provide a powerful tool for visualizing the lysosomal localization and catabolic degradation of antibodies and proteins. These kits utilize a unique fluorophore that is only active upon enzymatic cleavage by the lysosomal specific enzyme, Cathepsin B. Outside of the lysosome, LysoLight dyes have little to no observable background fluorescence, and only upon localization to the protease rich environment of the lysosome, does LysoLight become activated and brightly fluoresce bright green or deep red in color.
Specifications
Specifications
| Color | Verde |
| Método de detección | Fluorescente |
| Para utilizar con (aplicación) | Antibody and Protein Labeling |
| Para utilizar con (equipo) | Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument |
| Tipo de producto | Amine Reactive Dye |
| Características ecológicas | Enhanced stability |
| Línea de productos | LysoLight |
| Cantidad | 500 μg |
| Solubilidad | DMSO (dimetilsulfóxido) |
| Excitación/emisión | 488/525 nm |
Para uso exclusivo en investigación.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.