Learn More
Invitrogen™ LysoLight™ Antibody Labeling Kits
Descripción
LysoLight Antibody Labeling Kits provide a powerful tool for visualizing the lysosomal localization and catabolic degradation of antibodies and proteins. These kits utilize a unique fluorophore that is only active upon enzymatic cleavage by the lysosomal specific enzyme, Cathepsin B. Outside of the lysosome, LysoLight dyes have little to no observable background fluorescence, and only upon localization to the protease rich environment of the lysosome, does LysoLight become activated and brightly fluoresce bright green or deep red in color.
Especificaciones
Especificaciones
| Color | Verde |
| Método de detección | Fluorescente |
| Para utilizar con (aplicación) | Antibody and Protein Labeling |
| Para utilizar con (equipo) | Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument |
| Tipo de producto | Antibody Labeling Kit |
| Características ecológicas | Enhanced stability |
| Línea de productos | LysoLight |
| Cantidad | 3 x 1 mg |
| Solubilidad | DMSO (dimetilsulfóxido) |
| Excitación/emisión | 488/525 nm |
Para uso exclusivo en investigación.