Descripción
One-off maintenance service in accordance with maintenance schedule.
- Visual inspection of mechanical and electrical components, testing of all key functions.
- Calibration of a test temperature in the center of usable space as specified by the user
- Without certificate
Especificaciones
Especificaciones
| Para utilizar con (equipo) | Cámaras BINDER series KT, KB, KB ECO |
| Tipo de producto | Mantenimiento preventivo |