Learn More
MAP2K2 Mouse anti-Canine, Human, Mouse, Clone: OTI5B4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca:  Origene Technologies CF505762
Encompass_Preferred
Código de producto. 17624891
  • 750.20€ / 100 microgramos
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MEK1 (Mitogen activated protein kinase kinase 1) catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in MAP kinases. MEK1 activates ERK1 and ERK2 MAP kinases. Mitogen activated protein kinase kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.
Especificaciones
Especificaciones
MAP2K2
Monoclonal
Unconjugated
MAP2K2
CFC4, MAPKK2, MEK2, MKK2, PRKMK2
Mouse
Affinity Chromatography
RUO
26396, 5605, 611939
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunocytochemistry, Immunofluorescence, Western Blot
OTI5B4
PBS with 8% trehalose and no preservative; pH 7.3
P36507, Q63932
Map2k2
Full length human recombit protein of human MAP2K2 produced in HEK293T cell.
100 μg
Primary
Canine, Human, Mouse
Antibody
IgG1
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones