missing translation for 'onlineSavingsMsg'
Learn More

MATK Monoclonal Antibody (9D7), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA515553

Código de producto. 11395513

  • 461.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

MA5-15553 targets MATK in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-15553 immunogen is purified recombinant fragment of human MATK expressed in E. Coli. MA5-15553 detects MATK which has a predicted molecular weight of approximately 56kDa.

MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and/or differentiation. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

MATK
Monoclonal
1.0 mg/mL
PBS with 50% glycerol and 0.03% sodium azide
P42679
MATK
Purified recombinant fragment of human MATK expressed in E. Coli.
100 μL
Primary
Human
Antibody
IgG1
ELISA, Flow Cytometry, Western Blot
9D7
Unconjugated
MATK
Batk; CHK; Csk homologous kinase; Csk-homologous kinase; Csk-type protein tyrosine kinase; CTK; DKFZp434N1212; Hematopoietic consensus tyrosine-lacking kinase; HHYLTK; hydroxyaryl-protein kinase; HYL; HYL tyrosine kinase; HYLTK; leukocyte carboxyl-terminal src kinase related; Lsk; Matk; megakaryocyte-associated tyrosine kinase; megakaryocyte-associated tyrosine kinase (non-receptor protein tyrosine kinase); megakaryocyte-associated tyrosine-protein kinase; MGC1708; MGC2101; non-receptor protein kinase protein; Ntk; Protein kinase BATK; protein kinase HYL; Protein kinase NTK; tyrosine kinase MATK; tyrosine-protein kinase CTK; tyrosylprotein kinase
Mouse
Protein G
RUO
4145
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales