missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menaquinone-7, MedChemExpress
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
Description
Menaquinone-7 (Vitamin K2-7) is identified as the most bioactive cofactor for the carboxylation reaction of Gla-proteins . Supplementation with Menaquinone-7 (Vitamin K2-7) is a pharmacological option for activating matrix Gla protein and intervening in the progression of calcific aortic valve stenosis (CAVS).
Specifications
Specifications
| Nombre del producto químico o material | Menaquinone-7 |
| CAS | 2124-57-4 |
| Color | Yellow |
| Notas de grado de pureza | Research |
| Fórmula molecular | C46H64O2 |
| Cantidad | 5 mg |
| Sinónimo | Vitamin K2-7 Vitamin K2(35) Vitamin MK-7 |
| Información de solubilidad | DMSO : 5 mg/mL (7.70 mM; Need ultrasonic and warming) |
| SMILES | O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)=C(C)C(C2=C1C=CC=C2)=O |
| Peso molecular (g/mol) | 649.0 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction