missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Metal Lysing Tube, 2 mL, with 0.75 x 0.5 in. Grinding Cylinder

Stainless steel lysing matrix tube with the grinding cylinder is ideal for grinding, lysis, and homogenization of resistant samples.

184.00€ - 466.00€

Especificaciones

Tipo de esfera Stainless Steel
Tipo de muestra Seeds, Wood, Bark, Roots, Bone, Teeth, Tissues, Cartilage, Rocks, Minerals, Plastics, Composites
Incluye Cilindro de molienda de 1,9 x 1,27 cm (0,75 x 0,5 pulg)
Tipo de producto Tubo de lisado de metal
Ver más especificaciones

Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15808674
Ver documentos
MP Biomedicals™
116992002
2 paq.
184.00€
paquete de 2
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
15818674
Ver documentos
MP Biomedicals™
116992003
3 paq.
244.00€
paquete de 3
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
15828674
Ver documentos
MP Biomedicals™
116992006
6 paq.
466.00€
paquete de 6
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

This stainless steel lysing matrix tube with the grinding cylinder is ideal for grinding, lysis, and homogenization of resistant samples. Constructed from 308 SS, these tubes and grinding cylinders are tough enough to stand up to the most demanding mechanical punishment that can cause traditional thermoplastic tubes to crack. The tubes are machined from premium grade billet and deliver superior strength over less expensive production methods such as deep-drawn aluminum tubes. An oblique angle conical bottom provides a better impact surface than the rounded bottoms of deep-drawn tubes. The stainless steel threaded cap provides a leak-proof closure without the energy-robbing alternatives like plastic flange screw caps or rubber stoppers. A Teflon O-ring prevents leakage, and can be cleaned with detergent and/or autoclaving, or replaced entirely between samples. Machined knurls on the cap provide a firm grip to easy opening and closing.

Sample types include:

  • Tough seeds such as dried corn, soybeans, wheat, tomato, and Chile
  • Wood, bark, roots, animal claws and hooves
  • Bone, teeth, hair, fingernails, and non-biological substrates
  • Tough tissues, bone, cartilage, skin
  • Non-biological, rocks and minerals, plastics and composites, printed circuit boards, wood, and building materials
Application Notes
  • Dry grinding of very tough or hard samples in which generated heat can damage plastic tubes.
  • Cryogenic dry grinding where severe cold temperatures (dry ice or liquid nitrogen) can damage plastic tubes.
  • Milling or grinding non-biological samples where plastic contamination is of concern.
  • Sample processing with solvents or chemicals that are incompatible with plastics.
Especificaciones

Especificaciones

Stainless Steel
Seeds, Wood, Bark, Roots, Bone, Teeth, Tissues, Cartilage, Rocks, Minerals, Plastics, Composites
Cilindro de molienda de 1,9 x 1,27 cm (0,75 x 0,5 pulg)
Tubo de lisado de metal
Videos
SDS
Documentos

Documentos

Product Certifications

A menos que se especifique lo contrario, se utilizan únicamente para la investigación o la fabricación ulterior, no para uso humano directo.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado