missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Fusion Protein Antibody (hMPV24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-50505-0.025ml
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Metapneumovirus Fusion Protein Monoclonal antibody specifically detects Metapneumovirus Fusion Protein in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Especificaciones
| Metapneumovirus Fusion Protein | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 20 μg/mL | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| hMPV24 | |
| Unconjugated | |
| PBS | |
| Human MPV strain NCL-145 | |
| 0.025 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu