Learn More
Microbiologics™ Escherichia coli ATCC™ 8739™†
For culture purposes and QC applications
Especificaciones
Especie | Escherichia coli |
---|---|
ATCC® | 8739 |
Nivel de bioseguridad | 1 |
Certificaciones/conformidad | CE, online certificate of analysis |
Contenido y almacenamiento | De 2 °C a 8 °C |
Descripción
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single strain of Escherichia coli in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2, 6 or 10
- The most economical reference stock culture option; lyophilized pellet containing a single strain of microorganism
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 6 or 10 pellets
- ATCC™ 8739™†
Especificaciones
Escherichia coli | |
1 | |
De 2 °C a 8 °C | |
Culture purpose and QC application |
8739 | |
CE, online certificate of analysis | |
Lyophilized Pellet |
Seguridad y manipulación
missing translation for 'shelfLife' : 6 to 24 Months
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.