Learn More
Microbiologics™ EZ-CFU™ One-Step Microorganism Preparations
Lyophilized microorganism preparations deliver less than 100CFU per 0.1mL of hydrated suspension—no dilutions are required
650.79€
Especificaciones
Formato | EZ-CFU |
---|---|
Tipo de producto | EZ-CFU |
Includes: Product warranty and a peel-off Certificate of Assay
Código de producto | Marca | ATCC® | Formato | Especie | Precio | Cantidad y disponibilidad | |||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | ATCC® | Formato | Especie | Precio | Cantidad y disponibilidad | |||
11952745
|
Microbiologics™
0486Z |
6633 | EZ-CFU | Bacillus subtilis subsp. spizizenii |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||
Descripción
EZ-CFU One-Step preparations are designed to provide the required microorganism challenge concentrations for tests such as Growth Promotion Tests, Microbial Limits Tests and Microbial Enumeration Tests. These ready-to-use, dependable, and cost-effective enumerated microorganism preparations allow laboratories to conveniently and confidently conquer a variety of mandated Pharmacopoeial challenges.
- Up to 19 tests per 2mL aliquot or 190 total tests per kit can be completed
- Produces a durable, non-foaming suspension that is ready to be added to the challenge material immediately following the hydration step (30 minutes)
- For added convenience, a peel-off Certificate of Assay is included for quality control documentation
- Traceable to reference cultures
Información de órdenes
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Especificaciones
EZ-CFU | |
EZ-CFU |