missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, solución de tampón de 0,5 M, pH 8,0, Thermo Scientific™
Click to view available options
Cantidad:
100 mL
250 mL
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Nombre del producto químico o material | MOPS buffer soln. |
Concentración | Solución de tampón de 0,5 M |
pH | 8.0 |
Forma física | Líquida |
Cantidad | 100 mL |
Olor | Odorless |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'