Learn More
ESR1, Mouse, Clone: 5D4B1, 5D4E3, Abnova™
Mouse monoclonal antibody raised against partial recombinant ESR1.
Marca: Abnova MAB8304.100uL
Descripción
This gene encodes an estrogen receptor, a ligand-activated transcription factor composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5' UTRs and use different promoters. [provided by RefSeq]
Especificaciones
| ESR1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| ESR1 | |
| ESR1 | |
| Recombinant protein corresponding to truncated human ESR1. | |
| RUO | |
| 2099 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| ELISA, Western Blot | |
| 5D4B1, 5D4E3 | |
| Mouse monoclonal antibody raised against partial recombinant ESR1. | |
| In ascites | |
| DKFZp686N23123/ER/ESR/ESRA/Era/NR3A1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.