missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES1, Mouse, Clone: 3C7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HES1.
Marca: Abnova H00003280-M05A.200uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box. [provided by RefSeq
Sequence: APCKLGSQAGEAAKVFGGFQVVPAPDGQFAFLIPNGAFAHSGPVIPVYTSNSGTSVGPNAVSPSSGPSLTADSMWRPWRNEspecificaciones
HES1 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_005524 | |
HES1 | |
HES1 (NP_005515.1,201 a.a. ∽ 280 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
3280 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3C7 | |
Mouse monoclonal antibody raised against a partial recombinant HES1. | |
HES1 | |
FLJ20408/HES-1/HHL/HRY/bHLHb39 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
HES1, Mouse, Clone: 3C7, Abnova™ > 200μL; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido