Learn More
Invitrogen™ Mouse anti-Human IgG4 Fc Secondary Antibody, Biotin
Código de producto. 10365878
Change view
Click to view available options
Cantidad:
250 μg
Tamaño de la unidad:
cada uno
10365878 250 μg cada uno
1 options
Código de producto. 10365878
Marca: Invitrogen™ A10663
Mouse Monoclonal Secondary Antibody
Mouse anti-Human IgG4 Fc Secondary Antibody for ICC/IF, Western Blot
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Antígeno Human IgG4 Fc
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon HP6025
Concentración 1 mg/mL
Conjugado Biotin
Formulación PBS with 5mM sodium azide; pH 7.5
Especie del huésped Mouse
Inmunógeno Human IgG4.
Método de purificación Affinity chromatography
Cantidad 250 μg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Human
Contenido y almacenamiento 4°C
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG1