Learn More
NTRK1, Mouse, Clone: 6B2, Abnova™
Mouse monoclonal antibody raised against recombinant NTRK1.
Marca: Abnova MAB10431.100uL
Descripción
This gene encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of this kinase leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in this gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date. [provided by RefSeq
Especificaciones
NTRK1 | |
Monoclonal | |
Unconjugated | |
Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
NTRK1 | |
NTRK1 | |
Recombinant protein corresponding to human NTRK1. | |
RUO | |
4914 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunofluorescence, Western Blot | |
6B2 | |
Mouse monoclonal antibody raised against recombinant NTRK1. | |
In ascitic (0.03% sodium azide) | |
DKFZp781I14186/MTC/TRK/TRK1/TRKA/p140-TrkA | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.