Learn More
Mouse anti-Rat IgG2b Secondary Antibody, PE, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen SA510186
Detalles adicionales : Peso : 0.01000kg
Descripción
SA5-10186 targets Rat IgG2b in FACS applications and shows reactivity with Rat samples. The SA5-10186 immunogen is rat IgG.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Especificaciones
Rat IgG2b | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Rat IgG | |
50 μg | |
Secondary | |
4° C, do not freeze | |
IgG1 κ |
Flow Cytometry | |
NORIG 7.16.2 | |
PE | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Liquid |