Learn More
SOD1, Mouse, Clone: 6F5, Abnova™
Mouse monoclonal antibody raised against recombinant SOD1.
Marca: Abnova MAB10394.100uL
Descripción
The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq
Especificaciones
| SOD1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow cytometry (1:200-1:400) Immunofluorescence (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| SOD1 | |
| SOD1 | |
| Recombinant protein corresponding to full length human SOD1. | |
| RUO | |
| 6647 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Western Blot | |
| 6F5 | |
| Mouse monoclonal antibody raised against recombinant SOD1. | |
| In ascites (0.03% sodium azide) | |
| ALS/ALS1/IPOA/SOD/homodimer | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.