missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYK, Mouse, Clone: SYK-01, Abnova™
Mouse monoclonal antibody raised against partial recombinant SYK.
Marca: Abnova MAB0948.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
SYK | |
Monoclonal | |
1 mg/mL | |
Mouse monoclonal antibody raised against partial recombinant SYK. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
DKFZp313N1010/FLJ25043/FLJ37489 | |
Mouse | |
100 μg | |
Primary | |
This antibody reacts with protein tyrosine kinase p72Syk (Syk; Syk family tyrosine-specific phospho-transferase), which is required for the transduction of signals through the B cell antigen receptor (BCR) and the high affinity IgE receptor (Fc epsilon RI). | |
Store at 4°C. Do not freeze. | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
SYK-01 | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
SYK | |
SYK | |
Recombinant protein corresponding to amino acids 5-360 of human SYK. | |
RUO | |
6850 | |
Human, Mouse, Rat | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido