missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CCL17 (TARC) Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
500 microgramos
Descripción
Recombinant Murine TARC (CCL17) is an 7.9 kDa protein containing 70 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
Especificaciones
Especificaciones
| Para utilizar con (aplicación) | Functional Assay |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 20295 |
| Peso molecular | 7.9 kDa |
| Nombre | Mouse CCL17 (TARC) |
| Cantidad | 100 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Alias de gen | A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido