Learn More
Gibco™ Mouse CCL21 (6Ckine) Recombinant Protein
Beschreibung
Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute this protein in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Do not vortex. Further dilutions should be made in aqueous buffers supplemented with carrier protein, such as 0.1-1.0% BSA. Storage: This lyophilized preparation is stable at room temperature for 1 month, but should be kept at -20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at -20°C to -80°C for 3months. Avoid repeated freeze/thaw cycles.
Spezifikation
Spezifikation
| Número de acceso | P84444, P86792, P86793 |
| Para utilizar con (aplicación) | Bioactivity |
| Formulación | Protein with no preservative |
| ID de gen (Entrez) | 100042493, 18829, 65956 |
| Peso molecular | 12 kDa |
| Nombre | Mouse CCL21 (6Ckine) |
| Método de purificación | Purified |
| Cantidad | 20 μg |
| Requisitos de almacenamiento | -20°C |
| Estado normativo | RUO |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.