missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR8 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marca: R&D Systems MAB8324
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CCR8 Monoclonal specifically detects CCR8 in Mouse samples. It is validated for Flow Cytometry.Especificaciones
CCR8 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
1237 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Flow Cytometry | |
1055C | |
Flow Cytometry 0.25 ug/10^6 cells | |
P56484 | |
CCR8 | |
HEK293 human embryonic kidney cell line transfected with mouse CCR8 Accession # P56484 | |
100 μg | |
Primary | |
Stains mouse CCR8 transfectants but not irrelevant transfectants in flow cytometry. | |
Mouse | |
Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido