missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Dkk-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF2435-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Dkk-2 Polyclonal specifically detects Dkk-2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| Dkk-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, Dickkopf-2, dickkopf-related protein 2, Dkk2, DKK-2, hDkk-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27123 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9QYZ8 | |
| DKK2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Dkk-2 Ser26-Ile259 Accession # Q9QYZ8 | |
| 25 μg | |
| Primary | |
| Detects mouse Dkk-2 in Western blots. | |
| Mouse | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido