missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse Galectin-4 / LGALS4 / L36LBP Protein, His tag
495.95€ - 3302.36€
Especificaciones
Número de acceso | NP_034836.1 |
---|---|
Peso molecular | 38.6 kDa |
Fuente | Mouse Galectin-4, His Tag (GA4-M5247) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ile 326 (Accession # Q8K419-1). |
Inmunógeno | Met 1 - Ile 326 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15876807
|
ACROBiosystems
GA4-M5247-100UG |
100 μg |
495.95€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15886807
|
ACROBiosystems
GA4-M5247-1MG |
1 mg |
3302.36€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Galectin-4 (Gal-4), also known as L36LBP (L-36 lactose-binding protein). The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Galectin-4 expression is concentrated within microvilli in the gastrointestinal epithelium, where it can interact with CD3 and bind activated T cells in the lamina propria during intestinal inflammation. LGALS4 is an S-type lectin that is strongly underexpressed in colorectal cancer.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 38.6 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_034836.1 | |
Mouse Galectin-4, His Tag (GA4-M5247) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ile 326 (Accession # Q8K419-1). | |
-20°C | |
Galectin-4 | |
Mouse | |
50 mM HEPES, 150 mM NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
38.6 kDa | |
Met 1 - Ile 326 | |
LGALS4,L36LBP,Gal-4,Antigen NY-CO-27,Lactose-binding lectin 4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |