missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GFR alpha-like Antibody, R&D Systems™
Rat Monoclonal Antibody
Marca: R&D Systems MAB5728-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GFR alpha-like Monoclonal antibody specifically detects GFR alpha-like in Mouse samples. It is validated for ImmunocytochemistryEspecificaciones
GFR alpha-like | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Immunocytochemistry | |
1045708 | |
Immunocytochemistry 8-25 ug/mL | |
Q6SJE0-1 | |
Chinese Hamster Ovary cell line CHO-derived mouse GFR alpha-like, Gln20-Gly349, Accession # Q6SJE0-1 | |
25 μg | |
Primary | |
Mouse | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido