Learn More
Mouse IgG F(ab')2 Isotype Control, Invitrogen™
Mouse Isotype Control
Marca: Invitrogen 31203
Detalles adicionales : Peso : 0.80000kg
Descripción
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity: The precipitin lines against goat anti-mouse whole serum are the same as those against goat anti-mouse IgG, F(ab')2 fragment specific when assayed by immunoelectrophoresis at an antigen concentration of 20 mg/mL. No reaction was observed against goat anti-mouse IgG, Fc fragment specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Storage: After opening, dilute only enough antibody for a single day’s use. Country of Origin: USA
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody's species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.Especificaciones
Mouse IgG F(ab')2 | |
4.8 mg/mL | |
PBS with no preservative; pH 7.6 | |
Mouse | |
2 mg | |
Isotype Control (Primary) | |
4° C | |
IgG |
Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
Unconjugated | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; VH7183 | |
purified | |
RUO | |
Not Applicable | |
Liquid |