Learn More
Mouse IgG2a Isotype Control, Invitrogen™
Mouse Isotype Control
Marca: Invitrogen 026200
Detalles adicionales : Peso : 0.02100kg
Descripción
The protein concentration is 1 mg/mL based on E(1%) = 14 at 280 nm. This myeloma protein was purified from mouse ascites induced by myeloma cells. Specificity: This mouse myeloma IgG2a was characterized by enzyme immunoassay (ELISA).
Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Especificaciones
Mouse IgG2a | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Mouse | |
purified | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Control, ELISA | |
Unconjugated | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Mouse IgG2a | |
1 mg | |
Not Applicable | |
Liquid |