Learn More
ACROBiosystems Mouse IL-13 R alpha 1 Protein, His Tag
302.42€ - 1608.84€
Especificaciones
Número de acceso | NP_598751 |
---|---|
Peso molecular | 37.9 kDa |
Fuente | Mouse IL-13 R alpha 1, His Tag (IL1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession # NP_598751). |
Inmunógeno | Ala 26 - Thr 340 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15887027
|
ACROBiosystems
IL1-M52H7-1MG |
1 mg |
1608.84€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15877027
|
ACROBiosystems
IL1-M52H7-100UG |
100 μg |
302.42€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Interleukin 13 receptor, alpha 1 is also known as IL13RA1, NR4 and CD213A1 (cluster of differentiation 213A1), The IL13 Rα1 cDNA encodes a 427 amino acid (aa) residue precursor protein with a putative 21 aa residue signal peptide, a 324 aa residue extracellular domain, a 23 aa residue transmembrane region and a 59 aa residue cytoplasmic tail. Human and mouseIL13Rα1 share 76% aa sequence identity. IL13RA1 is a subunit of the interleukin 13 receptor. This subunit forms a receptor complex with IL4 receptor alpha, a subunit shared by IL13 and IL4 receptors. This subunit serves as a primary IL13-binding subunit of the IL13 receptor, and may also be a component of IL4 receptors. This protein has been shown to bind tyrosine kinase TYK2, and thus may mediate the signaling processes that lead to the activation of JAK1, STAT3 and STAT6 induced by IL13 and IL4.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_598751 | |
Mouse IL-13 R alpha 1, His Tag (IL1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession # NP_598751). | |
-20°C | |
IL-13 R alpha 1 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
37.9 kDa | |
Ala 26 - Thr 340 | |
IL13RA1,CD213A1,IL-13 Ra,NR4,RP13-128O4.2,IL13Rα1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |