missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MMP-12 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF3467
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MMP-12 Polyclonal specifically detects MMP-12 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Especificaciones
MMP-12 | |
Polyclonal | |
Unconjugated | |
EDL24933 | |
MMP12 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMP-12 Ala18-Cys462 Accession # EDL24933 | |
100 μg | |
Primary | |
Detects mouse MMP-12 in direct ELISAs and Western blots. In direct ELISAs,less than 20% cross-reactivity with recombinant human MMP-12 is observed, and less than 5% cross-reactivity with recombinant mouse (rm) MMP-3, rmMMP-8, and rmMMP-10 is observed. | |
Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP12, MMP-12 | |
Goat | |
Affinity Purified | |
RUO | |
4321 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mouse MMP-12 Antibody, R&D Systems™ > 100μg; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido