Learn More
STEMCELL Technologies Mouse Recombinant MIG (CXCL9)
Click to view available options
Cantidad:
25 μg
5 μg
Tamaño de la unidad:
25 microgramos
5 microgramos
Descripción
- Monokine induced by interferon-gamma (MIG), or CXCL9, is a member of the CXC chemokine family
- MIG is closely related to two other chemokines: CXCL10 and CXCL11, all of which signal through the CXCR3 receptor (Ding et al.) MIG is secreted by a variety of immune cells including T cells, NK cells, dendritic cells, macrophages, and eosinophils, as well as non-immune cells including hepatic stellate cells, preadipocytes, thyrocytes, endothelial cells, tumor cells, fibroblasts, and glial cells of the central nervous system
- MIG has also been shown to act as a chemoattractant for activated T cells and for tumor-infiltrating leukocytes (TILs), but not for neutrophils or for monocytes
- MIG has also been reported to be both a tumor suppressor and tumor promoter in various types of cancer
Especificaciones
Especificaciones
| Nombre | Chemokine (C-C-C motif) ligand 9, CMK, crg-10, CXCL9, Gamma interferon-induced monokine, Humig, M119, MIG, Monokine induced by gamma-interferon, SCYB9, Small inducible cytokine B9 |
| Cantidad | 5 μg |
| Tipo de célula | Monocytes; T Cells |
| Grado de pureza o calidad | ≥95% |