Learn More
R&D Systems™ Mouse TNF-alpha DuoSet ELISA
Mouse TNF-alpha DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marca: R&D Systems™ DY410-05
Descripción
The Mouse TNF-alpha DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TNF-alpha.
Especificaciones
ELISA Development Kit | |
Mouse | |
31.20 to 2000 pg/mL | |
ELISA | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
21926 | |
5 Plates |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.