missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGPRF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA132845
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PA1-32845 detects GPCR RTA from human samples.
May bind to a neuropeptide and may regulate nociceptor function and/or development, including the sensation or modulation of pain. Expressed in the Gut, vas deferens, uterus and aorta; barely detectable in liver, kidney, lung, and salivary gland. In the brain, markedly abundant in the cerebellum.Especificaciones
MRGPRF | |
Polyclonal | |
Unconjugated | |
MRGPRF | |
BC019711; G protein coupled receptor (RTA); G protein coupled receptor Rca; G protein-coupled receptor 140; G protein-coupled receptor 168; G protein-coupled receptor MrgF; G protein-coupled receptor RTA; G-p; GPR140; GPR168; G-protein coupled receptor 140; G-protein coupled receptor 168; G-protein coupled receptor RTA; MAS related GPR family member F; mas-related G protein-coupled MRGF; Mas-related G protein-coupled receptor c; mas-related gene; mas-related gene F protein; MAS-related GPR, member F; mas-related G-protein coupled receptor member F; MAS-related G-protein coupled receptor, member F; MGC21621; Mgrc; MRGF; MRGPRF; PSEC0142; Rta; seven transmembrane helix receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96AM1 | |
MRGPRF | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human MRGPRF | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |