missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Monoclonal Antibody (3E1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA517129
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-17129 targets MSH6 in FACS, ICC, IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17129 immunogen is purified recombinant fragment of human MSH6 expressed in E. Coli. MA5-17129 detects MSH6 which has a predicted molecular weight of approximately 160kDa.
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from μgtraviolet-induced malignant transformation and hence the prevention of skin cancer.Especificaciones
MSH6 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P52701 | |
MSH6 | |
Purified recombinant fragment of human MSH6 expressed in E. Coli. | |
RUO | |
2956 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3E1 | |
Unconjugated | |
MSH6 | |
AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |