missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYC(phospho T358) & MYC Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Marca: Abnova DP0144.1Set
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein phosphorylation antibody pair set comes with two antibodies, one against the MYC protein, and the other against the specific T358 phosphorylated site of MYC for use in in situ Proximity Ligation Assay.Especificaciones
| MYC | |
| Unconjugated | |
| bHLHe39/c-Myc | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-MYC T358 rabbit polyclonal antibody (20 ul) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. MYC mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| v-myc myelocytomatosis viral oncogene homolog (avian) | |
| MYC | |
| RUO | |
| 4609 | |
| Human | |
| Ab Pair |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido