missing translation for 'onlineSavingsMsg'
Learn More

n-Amyl Alcohol, Analytical Grade, Penta Chemicals

Código de producto. 30020664
Click to view available options
:
n-Amyl Alcohol
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30020664

Marca: PENTA CHEMICALS 2206011000

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Especificaciones

Cantidad 1 L
Peso molecular (g/mol) 88.15
Porcentaje de pureza ≥99%
Grado Analítica
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • n-Amyl alcohol
Palabra de advertencia
  • Peligro
Clasificación
  • Toxicidad aguda Categoría 4
  • Lesión ocular grave/irritación ocular Categoría 1
  • Líquido inflamable Categoría 3
  • Corrosión o irritación cutáneas Categoría 2
  • Toxicidad específica para determinados órganos Categoría 3
Indicaciones de peligro
  • H226-Líquidos y vapores inflamables.
  • H315-Provoca irritación cutánea.
  • H318-Provoca lesiones oculares graves.
  • H332-Nocivo en caso de inhalación.
  • H335-Puede irritar las vías respiratorias.
Consejos de prudencia
  • P261-Evitar respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
  • P302+P352-EN CASO DE CONTACTO CON LA PIEL: Lavar con abundante agua/.
  • P304+P340-EN CASO DE INHALACIÓN: Transportar a la persona al aire libre y mantenerla en una posición que le facilite la respiración.
  • P305+P351+P338-EN CASO DE CONTACTO CON LOS OJOS: Aclarar cuidadosamente con agua durante varios minutos. Quitar las lentes de contacto, si lleva y resulta fácil. Seguir aclarando.
Otros peligros
  • MIXTURE LIST-Contém: 1-pentanol
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado