missing translation for 'onlineSavingsMsg'
Learn More

Éster etílico de N-Boc-glicina, 95 %, Thermo Scientific Chemicals

Código de producto. 15409128
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 14719-37-0
Fórmula molecular C9H17NO4
Peso molecular (g/mol) 203.238
Número MDL MFCD05663720
Clave InChI CNIBHMMDDXGDNR-UHFFFAOYSA-N
Sinónimo n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
PubChem CID 357729
Nombre IUPAC etil 2-[(2-metilpropan-2-il)oxicarbonilamino]acetato
SMILES CCOC(=O)CNC(=O)OC(C)(C)C

Especificaciones

Nombre del producto químico o material N-Boc-glycine ethyl ester
Color Incolora
Fórmula lineal (CH3)3COCONHCH2CO2CH2CH3
Cantidad 5 g
Formula Weight (peso de la fórmula) 203.24
Porcentaje de pureza 95%
Forma física Líquido aceitoso

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'