Learn More
NAD 299 hydrochloride, Tocris Bioscience™
Código de producto. 18084432
Click to view available options
Cantidad:
10 mg
50 mg
Selective, high affinity 5-HT1A antagonist
Selective, high affinity 5-HT1A receptor antagonist (Ki = 0.6 nM in vitro). Enhances the action of selective 5-HT reuptake inhibitors and reverses citalopram -induced (Cat. No. 1427) inhibition of serotonergic cell firing.
CAS 184674-99-5
Fórmula molecular C18H24ClFN2O2
Peso molecular (g/mol) 354.85
Clave InChI GSZJANKLCPHEEX-BTQNPOSSSA-N
Sinónimo nad 299 hydrochloride, 3r-3-dicyclobutylamino-8-fluoro-3,4-dihydro-2h-1-benzopyran-5-carboxamide hydrochloride
PubChem CID 90488823
Nombre IUPAC (3R)-3-[di(cyclobutyl)amino]-8-fluoro-3,4-dihydro-2H-chromene-5-carboxamide;hydrochloride
SMILES C1CC(C1)N(C2CCC2)C3CC4=C(C=CC(=C4OC3)F)C(=O)N.Cl
Especificaciones
Nombre del producto químico o material NAD 299 hydrochloride
Cantidad 10 mg
Formula Weight (peso de la fórmula) 354.85
Porcentaje de pureza >98%