missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NanoLuc™ (Nluc) Luciferase Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB100261-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Luciferase Monoclonal specifically detects Luciferase in Multi-Species samples. It is validated for Western Blot.
Especificaciones
| Luciferase | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 965853 | |
| Western Blot 2 μg/mL | |
| ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
| Synthetic peptide for NanoLuc™ (Nluc) Luciferase | |
| 25 μg | |
| Detects NanoLuc™ (Nluc) Luciferase in direct ELISAs. In Western blots, no cross-reactivity with Large BiT is observed. | |
| Multi-species | |
| Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido