missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NCL 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Marca: Abnova™ H00004691-T02.100uL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
Western Blot | |
NCL 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 76.6kDa | |
Transient overexpression cell lysate was tested with anti-NCL antibody (Mfr. No. H00004691-D01P) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
NM_005381 | |
pCMV-NCL full-length | |
100 μL | |
Human |