Learn More
β-Nicotinamide mononucleotide, MedChemExpress
MedChemExpress β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.
Marca: Medchem Express HY-F0004
Descripción
β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.Especificaciones
β-Nicotinamide mononucleotide | |
White | |
C11H15N2O8P | |
β-NM NMN | |
NC(C1=C[N+]([C@@H]2O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]2O)=CC=C1)=O | |
334.22 | |
Research |
1094-61-7 | |
Research | |
100 mg | |
H2O : 83.33 mg/mL (249.33 mM; Need ultrasonic) | |
334.22 | |
98.0% | |
Solid |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Research purposes only