Learn More
Nkx2.5 Monoclonal Antibody (2E1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515551
Detalles adicionales : Peso : 0.01000kg
Descripción
MA5-15551 targets NKX2.5 in indirect ELISA and WB applications, this antibody also shows reactivity with Human samples. The MA5-15551 immunogen is purified recombinant fragment of human NKX2.5 expressed in E. Coli. MA5-15551 detects NKX2.5 which has a predicted molecular weight of approximately 35kDa
This gene encodes a homeobox-containing transcription factor. This transcription factor functions in heart formation and development. Mutations in this gene cause atrial septal defect with atrioventricular conduction defect, and also tetralogy of Fallot, which are both heart malformation diseases. Mutations in this gene can also cause congenital hypothyroidism non-goitrous type 5, a non-autoimmune condition. Alternative splicing results in multiple transcript variants.Especificaciones
Nkx2.5 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.03% sodium azide | |
P52952 | |
Nkx2-5 | |
Purified recombinant fragment of human NKX2.5 expressed in E. Coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
2E1 | |
Unconjugated | |
Nkx2-5 | |
Cardiac-specific homeobox; cardiac-specific homeobox 1; CHNG5; CSX; CSX1; Drosophila NK2 transcription factor related, locus 5; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; HLHS2; Homeobox protein CSX; homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; homeobox protein Nkx-2.5; NK2 homeobox 5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; NKX2.5; Nkx-2.5; Nkx-25; NKX2-5; NKX2E; NKX4-1; rNKx-2.5; tinman; tinman paralog; VSD3 | |
Mouse | |
Protein G | |
RUO | |
1482 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |