missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nkx2.5 Monoclonal Antibody (4B11), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA518671
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide Sequence: MFPSPALTPT PFSVKDILNL EQQQRSLAAA GELSARLEAT LAPSSCMLAA FKPEAYAGPE AAAPGLPELR AELGRAPSPA KCASAFPAAP AFYPRAYSDP DPAKDPRAEK KELCALQKAV ELEKTEADNA *
Nkx2.5 Monoclonal antibody specifically detects Nkx2.5 in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Especificaciones
Nkx2.5 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P52952 | |
Nkx2-5 | |
NKX2-5 (NP_004378, 1 a.a. ∽ 130 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunoprecipitation, Western Blot | |
4B11 | |
Unconjugated | |
Nkx2-5 | |
Cardiac-specific homeobox; cardiac-specific homeobox 1; CHNG5; CSX; CSX1; Drosophila NK2 transcription factor related, locus 5; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; HLHS2; Homeobox protein CSX; homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; homeobox protein Nkx-2.5; NK2 homeobox 5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; NKX2.5; Nkx-2.5; Nkx-25; NKX2-5; NKX2E; NKX4-1; rNKx-2.5; tinman; tinman paralog; VSD3 | |
Mouse | |
Affinity chromatography | |
RUO | |
1482 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |