Learn More
NLGN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551850
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: APAPTVNTHF GKLRGARVPL PSEILGPVDQ YLGVPYAAPP IGEKRFLPPE PPPSWSGIRN ATHFPPVCPQ NIHTAVPEVM LPVWFTANLD IVATYIQEPN EDCLYLNVYV PTED Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.Especificaciones
NLGN3 | |
Polyclonal | |
Unconjugated | |
NLGN3 | |
A230085M13Rik; ASPGX1; AUTSX1; Gliotactin homolog; HNL3; KIAA1480; neuroligin 3; neuroligin 3 protein; neuroligin-3; NL3; NLGN3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54413 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZ94 | |
NLGN3 | |
Recombinant protein corresponding to Human NLGN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only