missing translation for 'onlineSavingsMsg'
Learn More

NM23-H2/NME2 Antibody (2E3), PE, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-60241PE

Código de producto. 18691057

  • 551.00€ / 0.10 ml

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Description

Description

NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM23-H2/NME2
Monoclonal
PE
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunocytochemistry
2E3
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in
Recombinant human NM23-H2/NME2 (1-152aa) purified from E. coli
0.1 mL
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
4831
Store at 4C in the dark.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.