missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen 718600
Detalles adicionales : Peso : 0.50000kg
Descripción
NMDAR2B Polyclonal antibody specifically detects NMDAR2B in Rabbit, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Especificaciones
NMDAR2B | |
Polyclonal | |
Unconjugated | |
GRIN2B | |
AW490526; EIEE27; GluN2B; GluRepsilon2; glutamate [NMDA] receptor subunit epsilon-2; glutamate ionotropic receptor NMDA type subunit 2B; glutamate receptor; glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, NMDA2B; glutamate receptor, ionotropic, NMDA2B (epsilon 2); glutamate receptor, ionotropic, N-methyl D-aspartate 2B; GRIN 2B; GRIN2B; HGNC:4586; hNR 3; hNR3; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2B; MGC142178; MGC142180; MRD6; Nmdar2b; NMDE2; NME2; N-methyl D-aspartate receptor 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 2B; N-methyl-D-aspartate receptor subunit 3; NR2B; NR3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100353266, 24410 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q00960 | |
GRIN2B | |
Fusion protein including a 251 amino acid sequence from the N-terminal portion of the rat NMDAR 2B protein. | |
100 μg | |
Primary | |
Rabbit, Rat | |
Antibody | |
IgG |