missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMNAT1 Monoclonal Antibody (675112), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524304
Detalles adicionales : Peso : 0.00010kg
Descripción
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
NMNAT1 Monoclonal antibody specifically detects NMNAT1 in Human samples. It is validated for Immunohistochemistry (Frozen).Especificaciones
NMNAT1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9HAN9 | |
NMNAT1 | |
Sf 21-derived recombinant human NMNAT-1 Met1-Thr279 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Frozen) | |
675112 | |
Unconjugated | |
NMNAT1 | |
2610529L11Rik; 5730441G13Rik; D4Cole1e; deamido-NAD(+) diphosphorylase; deamido-NAD(+) pyrophosphorylase; EC 2.7.7.1; EC 2.7.7.18; id:ibd5068; im:7144541; LCA9; NAD(+) diphosphorylase; NAD(+) pyrophosphorylase; naMN adenylyltransferase 1; nicotinamide mononucleotide adenylyl transferase; nicotinamide mononucleotide adenylyl transferase 1; nicotinamide mononucleotide adenylyltransferase 1; nicotinamide mononucleotide adenylyltransferase 1-like protein; nicotinamide nucleotide adenylyltransferase 1; nicotinamide/nicotinate mononucleotide adenylyltransferase 1; nicotinamide/nicotinic acid mononucleotide adenylyltransferase 1; nicotinamide-nucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 1; NMN adenylyltransferase 1; NMN/NaMN adenylyltransferase 1; nmnat; NMNAT1; NMNATPNAT1; PNAT1; PNAT-1; pyridine nucleotide adenylyltransferase 1; zgc:110243 | |
Mouse | |
Protein A/G | |
RUO | |
64802 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |