missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nNOS Monoclonal Antibody (3G6B10), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen 372800
Detalles adicionales : Peso : 0.50000kg
Descripción
NOS activity requires homodimerization as well as three cosubstrates (L-arginine, NADPH and O2) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin and heme). Several distinct NOS isoforms have been described and been shown to represent the products of three distinct genes. These include two constitutive Ca2+/CaM-dependent forms of NOS, including NOS1 (also designated ncNOS) whose activity was first identified in neurons, and NOS3 (also designated ecNOS), first identified in endothelial cells.Especificaciones
nNOS | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P29475, P29476, Q9Z0J4 | |
NOS1 | |
Recombinant protein derived from the N-terminal region of rat nNOS protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
3G6B10 | |
Unconjugated | |
NOS1 | |
2310005C01Rik; bNOS; bNOS1; Brain NOS; Constitutive NOS; IHPS1; NC-NOS; neuronal nitric oxide synthase; neuronal nitric oxide synthase NOS1; neuronal NOS; nitric oxidase synthase; nitric oxide synthase; nitric oxide synthase 1; nitric oxide synthase 1 (neuronal); nitric oxide synthase 1, neuronal; nitric oxide synthase, brain; nNOS; N-NOS; nNOS; NOS type I; NO; NOS; NOS Type 1; NOS type I; Nos1; Nos-1; NOS-I; Peptidyl-cysteine S-nitrosylase NOS1; Phospho-bNOS; Phospho-Brain NOS | |
Mouse | |
Protein A | |
RUO | |
18125, 24598, 4842 | |
-20°C | |
Liquid |